Mouse IL-11
Supplier: Bandelin Electronic GmbH &Co.KG
010-001-V49-0010 010-001-V49-0100 010-001-V49-0002
75929-032EA 8070 USD InStock
75929-032 75929-036 75929-040 75929-044
Mouse IL-11
Proteins and Peptides
Recombinant Mouse IL-11 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin 11 (IL-11) is a pleotropic cytokine closely related to IL-6 in function, but is overall one of the lesser characterized interleukins. In hematopoietic cell populations, IL-11 is thought to induce megakaryocyte differentiation. In non-hematopoietic populations, IL-11 is thought to be able to stimulate hepatic acute-phase proteins. Human IL-11 can stimulate murine cells. Recombinant mouse IL-11 is a non-glycosylated protein, containing 179 amino acids, with a molecular weight of 19.3 kDa.
Interleukin-11 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-11 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty