Mouse Recombinant RELM alpha (from E. coli)
Supplier: Bandelin Electronic GmbH &Co.KG
010-001-W17-0100 010-001-W17-0025 010-001-W17-0005
75931-508EA 4582 USD InStock
75931-508 75931-510 75931-512 75931-514
Mouse Recombinant RELM alpha (from E. coli)
Proteins and Peptides
Recombinant Mouse RELM alpha control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Resistin-Like Molecule-alpha, or RELM-α, is a cytokine member to a unique family of proteins including Relm-β and Resistin. Relm-α and Resistin are secreted by adipocytes, where as Relm-β is secreted by intestinal epithelial cells. All members of the family share a high conserved C-terminal region with 10 Cysteine residues. Recombinant mouse RELM-α is a non-glycosylated protein, containing 88 amino acids, with a molecular weight of 10 kDa.
RELM alpha Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-RELM alpha in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty