Human TNF-alpha
Supplier: Bandelin Electronic GmbH &Co.KG
012-001-W31-0100 012-001-W31-0020 012-001-W31-0005
75933-216EA 4582 USD InStock
75933-216 75933-218 75933-220 75933-222
Human TNF-alpha
Proteins and Peptides
Recombinant Rat TNF-alpha control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Tumor Necrosis Factor alpha (TNFα) is an inflammatory cytokine secreted by macrophages, monocytes, neutrophils, T cells, NK-cells following their stimulation by bacterial LPS. TNFα activates signals through two receptors, TNF-R1, which is expressed on most cell types, and TNF-R2, which is expressed mainly on immune cells. TNFα can have many functions including, to stimulate of phagocytosis in macrophages, to chemoattract neutrophils, to increase insulin resistance and to induce fever. Recombinant rat TNFα is a non-glycosylated protein, containing 157 amino acids, with a molecular weight of 17.3 kDa.
Tumor Necrosis Factor alpha Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Tumor Necrosis Factor alpha in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty