Rat Blood In Heparin Sodium, Rockland Immunochemicals
Supplier: Rockland Immunochemical
R510-0100 R510-0050
75931-462EA 966.42 USD InStock
75931-462 75931-464
Rat Blood In Heparin Sodium, Rockland Immunochemicals
Blood
Rat Blood In Heparin Sodium
  • aseptically collected and prepared to order
  • blood draw in Anticoagulant
Rat whole blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Rat red blood cells are perishable and are collected and processed upon receipt of your order.
Washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty