Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-VISTA Armenian hamster Monoclonal Antibody [clone: 13F3]

Supplier: Bio X Cell
BP0310-50MG BP0310-25MG BP0310-5MG BP0310-100MG
103721-646EA 5100 USD InStock
103721-646 103721-648 103721-650 103721-652 76554-868 76556-536 76557-050 76557-252
Anti-VISTA Armenian hamster Monoclonal Antibody [clone: 13F3]
Antibodies
InVivoPlus anti-mouse VISTA, Clone: 13F3, Size: 100 mg, Isotype: Armenian hamster IgG, Applications: in vivo blocking of VISTA signaling, in vitro blocking of VISTA signaling, Flow cytometry

Type: Primary
Antigen: VISTA
Clonality: Monoclonal
Clone: 13F3
Conjugation:
Epitope:
Host: Armenian hamster
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty