Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper High quality, grains for elemental analyzers in inorganic materials

Supplier: Glassinstruments B.V.

338-353-00 338-353-12 338-353-10
101189-610EA 52.62 USD InStock
101189-610 101189-612 101189-524
Copper High quality, grains for elemental analyzers in inorganic materials
Copper


Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty