Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D(-)-Lysine monohydrochloride ≥99%, white to off-white powder

Supplier: A.W. SCHÜRR GmbH

00204-25G 00204-5G 00204-100G 00204-250G
102572-570EA 97.92 USD InStock
102572-570 102614-860 102631-374 102631-376
D(-)-Lysine monohydrochloride ≥99%, white to off-white powder
D(-)-Lysine monohydrochloride


Formula: C₆H₁₅ClN₂O₂
MW: 182.65 g/mol
Melting Pt: 266 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012920
CAS Number: 7274-88-6
Order Now
Specification Test Results
Heavy Metals ≤10 ppm
Optical Rotation –20 to –21.5º (C=8 in 6 N HCl)
Loss on Drying ≤0.5%
Residue On Ignition ≤0.5%
Assay ≥99%
Chiral Purity ≥99.5% (Assay, ee)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty