Wilkins Chalgren Anaerobic HiVegTM Agar/Broth Base, HiMedia Laboratories
Supplier: PAESEL+LOREI GMBH&CO
MV832-500G MV863-500G MV832-2.5KG MV832-5KG MV863-2.5KG MV863-5KG
61001-342EA 254.97 USD InStock
61001-342 61001-376 89229-190 89229-192 89229-262 89229-264
Wilkins Chalgren Anaerobic HiVegTM Agar/Broth Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Wilkins Chalgren Anaerobic HiVeg agar/broth base is used for the isolation and cultivation of anaerobic bacteria and also for susceptibility testing of anaerobic bacteria by the agar dilution method.
  • Appearance of powder: Yellow colored, slightly greenish tinge, homogeneous, free flowing powder
  • Colour and clarity: Medium amber colored, clear to slightly opalescent gel forms in petri plates, clear solution in tubes
Wilkins Chalgren HiVeg Media are prepared by replacing animal based peptones of bovine origin by vegetable peptones. The media have yeast extract to provide vitamins and other growth factors like purines and pyrimidines that are essential for the growth of P. melaninogenica. Arginine serves as an amino acid source especially for Eubacterium lentum and pyruvate is an energy source for Asaccharolytic cocci such as Veillonella. Pyruvate with arginine can provide proper environment for anaerobic growth. Ferric pyrophosphate is necessary for the growth of Bacteroides species and menadione for P. melaninogenica.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty