HL Extract Powder, HiMedia Laboratories
Supplier: PAESEL+LOREI GMBH&CO
RM326-500G RM326-10KG RM326-2.5KG RM326-25KG RM326-5KG
61001-614EA 231.85 USD InStock
61001-614 89229-878 89229-880 89229-882 89229-884
HL Extract Powder, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
HL extract powder is processed under controlled conditions to retain all the nutritive values of liver. It is incorporated in media which are used for the growth of fastidious anaerobic organisms.
  • Brownish yellow to brown homogenous free flowing powder, having characteristic odor but not putrescent
  • Freely soluble in distilled water, insoluble in alcohol and ether
HL extract powder can be used used for bulk production of vaccines, steroids and enzymes. It is equivalent to liver extract powder.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty