MicroMaxNS Disposable Protective Coveralls with Elastic Wirsts and Ankles, Attached Hood, and Zipper Closure, Lakeland Industries
Supplier: Lakeland
CTL428-LG CTL428-XL CTL428-2X CTL428-3X CTL428-4X CTL428-5X
76310-156CS 193.32 USD InStock
76310-156 76310-158 76310-160 76310-162 76310-164 76310-166 76310-168 76310-170
MicroMaxNS Disposable Protective Coveralls with Elastic Wirsts and Ankles, Attached Hood, and Zipper Closure, Lakeland Industries
Disposable Garments
These disposable protective coveralls are made from lightweight microporous fabric.
  • Microporous film laminated to polypropylene substrate
  • Serged seams
  • Blood-borne pathogen/viral barrier tested
MicroMax NS coverall offers the protection against dirt, dust, grease, grime and light chemical splash.
Attached hood with elastic wrists and ankles for better fit and increased protection.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty