PLET Agar Base, Modified, HiMedia
Supplier: PAESEL+LOREI GMBH&CO
M1451-500G M1451-2.5KG M1451-5KG
95021-146EA 192.19 USD InStock
95021-146 89224-054 89224-056
PLET Agar Base, Modified, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
PLET agar base medium is recommended for the selective isolation and cultivation of B. anthracis.
  • Appearance: Cream to yellow homogeneous free flowing powder
  • Gelling: Firm, comparable with 1.5% agar gel
  • Color and clarity of prepared medium: Light amber colored clear to slightly opalescent gel forms in petri plates
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty