Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-1-hydroxy-1H-benzotriazole ≥98% (by HPLC)

Supplier: A.W. SCHÜRR GmbH

16263-25G 16263-100G 16263-250G 16263-1KG
102567-494EA 160.77 USD InStock
102567-494 102595-780 102595-782 102608-142
6-Chloro-1-hydroxy-1H-benzotriazole ≥98% (by HPLC)
6-Chloro-1-hydroxy-1H-benzotriazole


Formula: C₆H₄ClN₃O
Storage Temperature: Refrigerator
MDL Number: MFCD00822645
CAS Number: 26198-19-6
Order Now
Specification Test Results
Appearance White to Off-White Crystalline Powder
Purity by HPLC ≥98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty