Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromocaproic acid ≥98%

Supplier: A.W. SCHÜRR GmbH
Danger

24477-250G 24477-1KG 24477-100G 24477-25G
102554-296EA 219.23 USD InStock
102554-296 102564-096 102569-286 102601-530
6-Bromocaproic acid ≥98%
6-Bromocaproic acid


Formula: BrH₂C(CH₂)₄COOH
MW: 195.06 g/mol
Boiling Pt: 165
Flash Pt: 95 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00004422
CAS Number: 4224-70-8
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Appearance Colorless Liquid or White Solid
Water Content (Karl Fisher) ≤0.5%
Purity ≥98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty