Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-FGA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Innovative Research Inc
ASMFBGNFITC10 ASMFBGNFITC1MG
103386-396EA 3987.69 USD InStock
103386-396 103387-978
Anti-FGA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Primary
Antigen: FGA
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty