Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-HRP Chicken Antibody (HRP (Horseradish Peroxidase))

Supplier: Innovative Research Inc
AVI-HRP-0.01ML AVI-HRP-0.05ML
103384-756EA 43.85 USD InStock
103384-756 103386-498
Anti-HRP Chicken Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: HRP
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty