Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-F5 Mouse Monoclonal Antibody [clone: 14G5-G11]

Supplier: Innovative Research Inc
FV14G5-0.1MG FV14G5-1MG
103384-910EA 246.15 USD InStock
103384-910 103386-620
Anti-F5 Mouse Monoclonal Antibody [clone: 14G5-G11]
Antibodies
Type: Primary
Antigen: F5
Clonality: Monoclonal
Clone: 14G5-G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty