Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-F8 Mouse Monoclonal Antibody [clone: 1C2-G10F10]

Supplier: Innovative Research Inc
FVIII1C2-1MG FVIII1C2-0.1MG
103386-634EA 1889.23 USD InStock
103386-634 103388-008
Anti-F8 Mouse Monoclonal Antibody [clone: 1C2-G10F10]
Antibodies
Type: Primary
Antigen: F8
Clonality: Monoclonal
Clone: 1C2-G10F10
Conjugation: Unconjugated
Epitope: Heavy Chain
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty