Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-F11 Mouse Monoclonal Antibody [clone: 14F8-F10]

Supplier: Innovative Research Inc
FXI14F8-0.1MG FXI14F8-1MG
103384-916EA 246.15 USD InStock
103384-916 103386-642
Anti-F11 Mouse Monoclonal Antibody [clone: 14F8-F10]
Antibodies
Type: Primary
Antigen: F11
Clonality: Monoclonal
Clone: 14F8-F10
Conjugation: Unconjugated
Epitope: Light Chain
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty