Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-F11 Mouse Monoclonal Antibody [clone: 9C9-E10F7]

Supplier: Innovative Research Inc
FXI9C9E100.1MG FXI9C9E10-1MG
103384-920EA 375.38 USD InStock
103384-920 103386-646
Anti-F11 Mouse Monoclonal Antibody [clone: 9C9-E10F7]
Antibodies
Type: Primary
Antigen: F11
Clonality: Monoclonal
Clone: 9C9-E10F7
Conjugation: Unconjugated
Epitope: Heavy Chain
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty