Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-F12 Mouse Monoclonal Antibody [clone: 20C8-6F9F1]

Supplier: Innovative Research Inc
FXII20C8-1MG FXII20C8-0.1MG
103386-658EA 1889.23 USD InStock
103386-658 103388-016
Anti-F12 Mouse Monoclonal Antibody [clone: 20C8-6F9F1]
Antibodies
Type: Primary
Antigen: F12
Clonality: Monoclonal
Clone: 20C8-6F9F1
Conjugation: Unconjugated
Epitope: Heavy Chain
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty