Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-PLAT Mouse Monoclonal Antibody [clone: H27B6]

Supplier: Innovative Research Inc
H27B6-1MG H27B6-10MG
103384-976EA 869.62 USD InStock
103384-976 103386-718
Anti-PLAT Mouse Monoclonal Antibody [clone: H27B6]
Antibodies
Type: Primary
Antigen: PLAT
Clonality: Monoclonal
Clone: H27B6
Conjugation: Unconjugated
Epitope: Light Chain
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Mouse;Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty