Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgG Fab Human Antibody

Supplier: Innovative Research Inc
HU-FAB-5MG HU-FAB-10MG
103385-296EA 438.46 USD InStock
103385-296 103386-990
Anti-IgG Fab Human Antibody
Antibodies
Type: Control
Antigen: IgG Fab
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG Fab
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty