Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IGHA1 Human Antibody

Supplier: Innovative Research Inc
HU-IGA1M-5MG HU-IGA1M-10MG
103385-306EA 423.85 USD InStock
103385-306 103387-000
Anti-IGHA1 Human Antibody
Antibodies
Type: Control
Antigen: IGHA1
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgA1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty