Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IGHA2 Human Antibody

Supplier: Innovative Research Inc
HU-IGA2M-5MG HU-IGA2M-10MG
103385-308EA 511.54 USD InStock
103385-308 103387-002
Anti-IGHA2 Human Antibody
Antibodies
Type: Control
Antigen: IGHA2
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgA2
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty