Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IGHG1 Human Antibody

Supplier: Innovative Research Inc
HU-IGG1-1MG HU-IGG1-5MG
103385-322EA 613.85 USD InStock
103385-322 103387-016
Anti-IGHG1 Human Antibody
Antibodies
Type: Control
Antigen: IGHG1
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty