Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-PLAU Mouse Monoclonal Antibody [clone: 724-3A1-8] (FITC (Fluorescein Isothiocyanate))

Supplier: Innovative Research Inc
HUPA3A108FIT.1 HUPA3A108FITC1
103385-346EA 375.38 USD InStock
103385-346 103388-348
Anti-PLAU Mouse Monoclonal Antibody [clone: 724-3A1-8] (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Primary
Antigen: PLAU
Clonality: Monoclonal
Clone: 724-3A1-8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty