Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-KNG1 Mouse Monoclonal Antibody [clone: 16E4-H1]

Supplier: Innovative Research Inc
KNG16E4-0.1MG KNG16E4-1MG
103385-398EA 246.15 USD InStock
103385-398 103387-068
Anti-KNG1 Mouse Monoclonal Antibody [clone: 16E4-H1]
Antibodies
Type: Primary
Antigen: KNG1
Clonality: Monoclonal
Clone: 16E4-H1
Conjugation: Unconjugated
Epitope: Light Chain
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty