Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-SERPINE1 Mouse Monoclonal Antibody [clone: 124K1]

Supplier: Innovative Research Inc
MA-124K1-10MG MA-124K1-1MG
103387-084EA 11619.23 USD InStock
103387-084 103388-042
Anti-SERPINE1 Mouse Monoclonal Antibody [clone: 124K1]
Antibodies
Type: Primary
Antigen: SERPINE1
Clonality: Monoclonal
Clone: 124K1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Rat;Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty