Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-LRP1 Mouse Monoclonal Antibody [clone: 5A6] (FITC (Fluorescein Isothiocyanate))

Supplier: Innovative Research Inc
MA5A6FITC1MG MA5A6FITC0.1MG
103387-110EA 2270.77 USD InStock
103387-110 103388-068
Anti-LRP1 Mouse Monoclonal Antibody [clone: 5A6] (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Primary
Antigen: LRP1
Clonality: Monoclonal
Clone: 5A6
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human;Mouse;Rabbit

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty