Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-F9 Rat Monoclonal Antibody [clone: 9041]

Supplier: Innovative Research Inc
MAMFIX10.1MG MAMFIX10.5MG
103385-414EA 358.08 USD InStock
103385-414 103388-360
Anti-F9 Rat Monoclonal Antibody [clone: 9041]
Antibodies
Type: Primary
Antigen: F9
Clonality: Monoclonal
Clone: 9041
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty