Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-SERPINE1 Mouse Monoclonal Antibody [clone: 671-3E5-29]

Supplier: Innovative Research Inc
PAI3E529-1MG PAI3E529-10MG
103385-602EA 578.46 USD InStock
103385-602 103388-718
Anti-SERPINE1 Mouse Monoclonal Antibody [clone: 671-3E5-29]
Antibodies
Type: Primary
Antigen: SERPINE1
Clonality: Monoclonal
Clone: 671-3E5-29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Rabbit;Pig

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty