Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-PLG Mouse Monoclonal Antibody [clone: 4G6-B11]

Supplier: Innovative Research Inc
PLG4G6B11-10MG PLG4G6B11-1MG
103387-364EA 4984.62 USD InStock
103387-364 103388-588
Anti-PLG Mouse Monoclonal Antibody [clone: 4G6-B11]
Antibodies
Type: Primary
Antigen: PLG
Clonality: Monoclonal
Clone: 4G6-B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty