Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-REN Mouse Monoclonal Antibody [clone: 4B5-E3] (Biotin)

Supplier: Innovative Research Inc
PREN4B5E3BIO.1 PREN4B5E3BIO1
103385-652EA 529.23 USD InStock
103385-652 103387-394
Anti-REN Mouse Monoclonal Antibody [clone: 4B5-E3] (Biotin)
Antibodies
Type: Primary
Antigen: REN
Clonality: Monoclonal
Clone: 4B5-E3
Conjugation: Biotin
Epitope: 30—43
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty