Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgG Rhesus monkey Polyclonal Antibody

Supplier: Innovative Research Inc
RS-GF-10MG RS-GF-50MG
103385-806EA 246.15 USD InStock
103385-806 103387-556
Anti-IgG Rhesus monkey Polyclonal Antibody
Antibodies
Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rhesus monkey
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty