Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-PLAU Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Innovative Research Inc
SASDUPAFITC1MG SASDUPAFITC10
103388-214EA 480 USD InStock
103388-214 103388-766
Anti-PLAU Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Primary
Antigen: PLAU
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Dog

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty