Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-KLK3 Sheep Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
SASHPSABIO1MG SASHPSABIO10MG
103385-936EA 344.62 USD InStock
103385-936 103387-622
Anti-KLK3 Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: KLK3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty