Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-SERPINA10 Sheep Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
SASHZPIBIO1MG SASHZPIBIO10MG
103385-994EA 436.92 USD InStock
103385-994 103387-696
Anti-SERPINA10 Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: SERPINA10
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty