Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-F9 Sheep Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
SASMFIXBIO1MG SASMFIXBIO10MG
103386-004EA 436.92 USD InStock
103386-004 103387-706
Anti-F9 Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: F9
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty