Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-SERPINI1 Sheep Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
SASMNSPHTBIO.5 SASMNSPHTBIO5
103386-036EA 627.69 USD InStock
103386-036 103388-808
Anti-SERPINI1 Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: SERPINI1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty