Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-PLAU Sheep Polyclonal Antibody

Supplier: Innovative Research Inc
SASRBUPA-1ML SASRBUPA-10ML
103386-122EA 412.31 USD InStock
103386-122 103387-800
Anti-PLAU Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PLAU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty