XLD HiVeg™ Agar, Himedia
Supplier: PAESEL+LOREI GMBH&CO
MV031-500G MV031-2.5KG MV031-5KG
95044-414EA 192.08 USD InStock
95044-414 89227-256 89227-258
XLD HiVeg™ Agar, Himedia
Media for Microbiology Dehydrated Media for Microbiology
Recommended for the selective isolation and enumeration of Salmonella serotype Typhi and other Salmonella species.
  • Can be used as a diagnostic aid in the identification of Enterobacteriaceae
Ideal medium for screening samples containing mixed flora of enteric pathogens as recovery of Salmonellae and Shigellae is not conspicuous by even profuse growth of other species (2, 3).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty