GenTegra® RNA Storage Solutions, NBS Scientific
Supplier: Nova Biostorage Plus
NBGTR1009 NBGTR1001
103289-510PK 2429 USD InStock
103289-510 103289-512 103289-514 103289-576 103289-578
GenTegra® RNA Storage Solutions, NBS Scientific
Nucleic Acid Storage Systems
Stabilize RNA at ambient temperature for storage, transport and every day handling.
  • Simple rehydration process for 100% sample recovery for downstream applications
GenTegra RNA protects RNA immediately from hydrolysis and oxidation, stabilizing it in liquid form or after drying for long-term ambient temperature storage, providing better protection from freezing which only slows those processes.
RNA samples stored in GenTegra RNA show no degradation after the equivalent of 4 years of storage at ambient temperature (via accelerated aging studies).
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty