Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-FKBP38 Mouse Monoclonal Antibody [clone: 391505]

Supplier: Novus Biologicals
MAB3580-SP MAB3580
103297-548EA 215.48 USD InStock
103297-548 103353-084
Anti-FKBP38 Mouse Monoclonal Antibody [clone: 391505]
Antibodies
FK506 Binding Proteins (FKBPs) are intracellular receptors for the immuno-suppressive drug FK506. The FKBP/FK506 complex exerts its immunosuppressive effects by inhibiting calcineurin, a calcium- and calmodulin-dependent serine/threonine phosphatase that functions as a critical signaling molecule during T-cell activation.

  • Reconstitute at 0.5 mg/ml in sterile PBS
  • Dilute 0.5 μg/ml for Western blot

It binds to and inhibits calcineurin even in the absence of FK506, indicating that FKBP38 is a constitutively active inhibitor of calcineurin. Additionally, FKBP38 co‑immunoprecipitates with Bcl-2 and Bcl-xL, suggesting that FKBP38 may regulate apoptosis by anchoring Bcl‑2 and Bcl‑xL to mitochondrial membranes.

Precaución: This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Type: Primary
Antigen: FKBP38
Clonality: monoclonal
Clone: 391505
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty