ChemMax® 2 Chemical Protective Coverall with Respirator-Fit Hood, Elastic Wrists and Ankles, Zipper Closure with Storm Flap, Lakeland Industries
Supplier: Lakeland
C2T132-2X C2T132-3X C2T132-4X C2T132-5X C2T132-LG C2T132-MD C2T132-XL
76314-042CS 332.1 USD InStock
76314-042 76314-080 76314-082 76314-084 76314-086 76314-088 76314-090
ChemMax® 2 Chemical Protective Coverall with Respirator-Fit Hood, Elastic Wrists and Ankles, Zipper Closure with Storm Flap, Lakeland Industries
Disposable Garments
Chemical protective coveralls are made with the proven protection of Saranex® 23P barrier film for protection in Hazmat, chemical handling, and environmental cleanup applications.
  • Heat sealed seams
  • Respirator-fit hood
ChemMax® 2 chemical protectve coveralls provide protection from chemical mixing and splash,environmental cleanup, and hazardous materials remediation. These garments feature an attached respirator-fit hood and heat sealed seams to ensure maximum protection.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty