ChemMax® 3 Chemical Protective Coverall with Respirator-Fit Hood, Elastic Wrists and Ankles, Zipper Closure with Storm Flap, Lakeland Industries
Supplier: Lakeland
C3T132-2X C3T132-3X C3T132-4X C3T132-5X C3T132-LG C3T132-MD C3T132-XL
76314-126CS 499.01 USD InStock
76314-126 76314-128 76314-130 76314-132 76314-134 76314-136 76314-138
ChemMax® 3 Chemical Protective Coverall with Respirator-Fit Hood, Elastic Wrists and Ankles, Zipper Closure with Storm Flap, Lakeland Industries
Disposable Garments
Chemical protective coveralls are made with advanced ChemMax® 3 chemcial protective fabric for proven protection against a broad spectrum of toxic industrial chemicals, chemical warfare agents, ammonia, and fentanyl.
  • ChemMax® 3 chemical protective film
  • Heat sealed seams
  • Respirator-fit hood
  • Elastic wrists and ankles
  • Exceptional protection from Ammonia and Fentanyl
  • Supported by Lakeland's PermaSURE® Chemical Toxicity Risk Modeling Software
ChemMax® 3 chemical protective coveralls provide protection from the most toxic industrial chemicals, chemical warfare agents. Heat sealed seams and a storm flap over the zipper ensure maximum protection.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty