SHOWA 240 Cut Resistant, Flame Retardant Gloves, Showa
Supplier: Showa
240-07 240-08 240-09 240-10 240-11
76314-946DZ 345.17 USD InStock
76314-946 76314-948 76314-950 76314-952 76314-954
SHOWA 240 Cut Resistant, Flame Retardant Gloves, Showa
Gloves
Forged with flame-resistant materials, the SHOWA 240 glove line features a 13 gauge knitted liner containing Kevlar®, Modacrylic, and Fibreglass, making the glove extremely strong and resilient.
  • Protects against arc flash: Arc flash level 2
  • Flame resistant materials
Flat dipped sponge neoprene coating provides excellent grip, and a seamless liner prevents irritation. The anatomical design helps prevent hand fatigue, while the coloring helps to extend wearable life. They offer a high resistance to cuts and reliable wrist protection.
SHOWA 240 series gloves are ideally suited for electrical maintenance, fracking, metal/wire handling, warm/hot component handling, and resistance welding applications in construction, electric, gas, manufacturing, metallurgy, mining, oil, and gas industries.
Certificaciones: CE marked; EN 388 Cut level 5 resistance; EN 407 Level 4 resistance to flammability.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty