Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium sulfate ≥99.0%, Reagent Grade

Supplier: BURDICK & JACKSON FKA SIGMA

A5132-1KG A5132-500G A5132-5KG
BJA5132-1KGEA 106.43 USD InStock
BJA5132-1KG BJA5132-500G BJA5132-5KG
Ammonium sulfate ≥99.0%, Reagent Grade
Ammonium sulphate


Formula: (NH₄)₂SO₄
Melting Pt: 350 °C
Density: 1.760 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
Order Now
Specification Test Results
Assay 99.0 - 101.0 %
Colour of the solution complying
Clearness of the solution complying
Iron (Fe) Max. 10 ppm
Lead (Pb) Max. 3 ppm
Selenium (Se) Max. 5 ppm

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty