Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium carbonate 98.5-100.5% (by anhydrous basis), Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP ,E170, precipitated, Fluka™

Supplier: BURDICK & JACKSON FKA SIGMA

12010-6X1KG 12010-2.5KG
BJ12010-1KGCS 330.92 USD InStock
BJ12010-1KG BJ12010-2.5KG
Calcium carbonate 98.5-100.5% (by anhydrous basis), Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP ,E170, precipitated, Fluka™
Calcium carbonate


Formula: CaCO₃
MW: 100.09 g/mol
Melting Pt: 825 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658
Order Now
Specification Test Results
Assay (calculated to the dried substance) 98.5 - 100.5 %
Insoluble in acetic acid Max. 0.2 %
Insoluble in hydrochloric acid Max. 0.2 %
Loss on drying (200°C) Max. 2.0 %
Magnesium and alkali metals Max. 1.0 %
Arsenic (As) Max. 2 ppm
Barium (Ba) Max. 10 ppm
Cadmium (Cd) Max. 1 ppm
Chromium (Cr) Max. 10 ppm
Copper (Cu) Max. 10 ppm
Iron (Fe) Max. 200 ppm
Mercury (Hg) Max. 0.5 ppm
Lead (Pb) Max. 3 ppm
Antimony (Sb) Max. 10 ppm
Zinc (Zn) Max. 10 ppm
Heavy metals (as Pb) Max. 20 ppm
Sb, Cu, Cr, Zn, Ba Max. 100 ppm
Chloride (Cl) Max. 200 ppm
Fluoride (F) Max. 50 ppm
Sulfate (SO₄) Max. 0.2 %
Residual solvents complying

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty