Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disodium hydrogen phosphate, anhydrous ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™

Supplier: BURDICK & JACKSON FKA SIGMA

71640-1KG 71640-250G 71640-5KG
BJ71640-1KGEA 147.04 USD InStock
BJ71640-1KG BJ71640-250G BJ71640-5KG
Disodium hydrogen phosphate, anhydrous ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Disodium hydrogen phosphate


Formula: Na₂HPO₄
MW: 141.96 g/mol
Melting Pt: 250
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
EINECS: 231-448-7
Merck Index: 12,08805
Order Now
Specification Test Results
Assay 99.0 - 101.0 %
Colour of the solution complying
Clearness of the solution complying
pH (5 %, 25 °C) 8.7 - 9.3
Loss on drying (105°C) Max. 0.2 %
Water insoluble matter Max. 0.01 %
Arsenic (As) Max. 1 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 10 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 2 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 10 ppm
Total S (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 10 ppm
Total N Max. 10 ppm

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty