Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium chloride ≥99.5% (by argentometric titration), Puriss. p.a.

Supplier: BURDICK & JACKSON FKA SIGMA

60130-1KG 60130-250G 60130-5KG
BJ60130-1KGEA 171.69 USD InStock
BJ60130-1KG BJ60130-250G BJ60130-5KG
Potassium chloride ≥99.5% (by argentometric titration), Puriss. p.a.
Potassium chloride


Formula: KCl
Boiling Pt: 1.413 °C (1.013 hPa)
Melting Pt: 772 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Order Now
Specification Test Results
Assay (argentometric) 99.5 - 101.0 %
pH (5 %, 20°C) 5.5 - 8.0
Calcium (Ca) Max. 0.005 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Chromium (Cr) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0002 %
Magnesium (Mg) Max. 0.001 %
Manganese (Mn) Max. 0.0005 %
Sodium (Na) Max. 0.02 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Bromide (Br) Max. 0.05 %
Iodide (I) Max. 0.002 %
Phosphate (PO₄) Max. 0.0005 %
Sulfate (SO₄) Max. 0.003 %
Total N Max. 0.001 %
Colour of the solution complying
Turbidity of solution complying

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty